2-Fluoro-6-methyl-3-pyridinecarboxylic acid ≥95%

Supplier: AOBChem USA

Warning

21665-1G 21665-500MG 21665-5G 21665-25G
77466-948EA 120.51 USD
77466-948 77479-678 77489-966 77500-222
2-Fluoro-6-methyl-3-pyridinecarboxylic acid ≥95%
2-Fluoro-6-methyl-3-pyridinecarboxylic acid

Precaución: For research use only

Formula: C₇H₆FNO₂
MW: 155.126 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16611276
CAS Number: 1211583-88-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR