2-(2,2,2-Trifluoroethoxy)-6-(trifluoromethyl)-3-pyridinecarboxylic acid ≥95%

Supplier: AOBChem USA

Warning

215889-1G 215889-500MG 215889-5G
77466-944EA 574.74 USD
77466-944 77479-674 77489-962
2-(2,2,2-Trifluoroethoxy)-6-(trifluoromethyl)-3-pyridinecarboxylic acid ≥95%
2-(2,2,2-Trifluoroethoxy)-6-(trifluoromethyl)-3-pyridinecarboxylic acid

Precaución: For research use only

Formula: C₉H₅F₆NO₃
Storage Temperature: Ambient
MDL Number: MFCD22888788
CAS Number: 1505575-72-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR