Methyl 5-fluoro-2-(trifluoromethyl)isonicotinate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Methyl 5-fluoro-2-(trifluoroMethyl)-4-pyridinecarboxylate, Methyl 5-fluoro-2-(trifluoromethyl)pyridine-4-carboxylate

215705-1G 215705-500MG 215705-5G
77466-940EA 447.02 USD
77466-940 77479-670 77489-958
Methyl 5-fluoro-2-(trifluoromethyl)isonicotinate ≥97%
Methyl 5-fluoro-2-(trifluoromethyl)isonicotinate

Precaución: For research use only

Formula: C₈H₅F₄NO₂
MW: 223.124 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28799126
CAS Number: 1803834-94-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR