2-Methyl-5-propoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

21548-1G 21548-500MG 21548-5G 21548-25G
77466-936EA 397.58 USD
77466-936 77479-666 77489-954 77500-212
2-Methyl-5-propoxyphenylboronic acid ≥97%
2-Methyl-5-propoxyphenylboronic acid

Precaución: For research use only

Formula: C₁₀H₁₅BO₃
MW: 194.035 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19105380
CAS Number: 1451391-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR