α-Butyl-2-chloro-6-fluorobenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

214659-1G 214659-500MG 214659-5G 214659-25G
77466-932EA 353.29 USD
77466-932 77479-664 77489-950 77500-206
α-Butyl-2-chloro-6-fluorobenzenemethanol ≥95%
α-Butyl-2-chloro-6-fluorobenzenemethanol

Precaución: For research use only

Formula: C₁₁H₁₄ClFO
MW: 216.68 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21140792
CAS Number: 1494885-59-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR