α-Butyl-4-(ethylthio)benzenemethanol ≥97%

Supplier: AOBChem USA

Warning

214596-1G 214596-500MG 214596-5G 214596-25G
77466-924EA 312.09 USD
77466-924 77479-656 77489-942 77500-198
α-Butyl-4-(ethylthio)benzenemethanol ≥97%
α-Butyl-4-(ethylthio)benzenemethanol

Precaución: For research use only

Formula: C₁₃H₂₀OS
MW: 224.36 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34867694
CAS Number: 1693894-21-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR