α-Butyl-4-(2-methylpropyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

214538-1G 214538-500MG 214538-5G 214538-25G
77466-920EA 415.09 USD
77466-920 77479-652 77489-938 77500-194
α-Butyl-4-(2-methylpropyl)benzenemethanol ≥95%
α-Butyl-4-(2-methylpropyl)benzenemethanol

Precaución: For research use only

Formula: C₁₅H₂₄O
MW: 220.35 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12786725
CAS Number: 148367-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR