α-Butyl[1,1'-biphenyl]-4-methanol ≥95%

Supplier: AOBChem USA

Warning

214508-1G 214508-500MG 214508-5G
77466-916EA 487.19 USD
77466-916 77479-648 77489-934
α-Butyl[1,1'-biphenyl]-4-methanol ≥95%
α-Butyl[1,1'-biphenyl]-4-methanol

Precaución: For research use only

Formula: C₁₇H₂₀O
MW: 240.34 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16357168
CAS Number: 50673-89-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR