4-Butyl-α-(1-methylpropyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

214457-1G 214457-500MG 214457-5G 214457-25G
77466-912EA 415.09 USD
77466-912 77479-644 77489-930 77500-190
4-Butyl-α-(1-methylpropyl)benzenemethanol ≥95%
4-Butyl-α-(1-methylpropyl)benzenemethanol

Precaución: For research use only

Formula: C₁₅H₂₄O
MW: 220.35 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12787350
CAS Number: 1248314-12-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR