2,4,6-Trifluoro-α-(1-methylpropyl)benzenemethanol ≥97%

Supplier: AOBChem USA

Warning

214442-1G 214442-500MG 214442-5G 214442-25G
77466-908EA 255.44 USD
77466-908 77479-640 77489-926 77500-186
2,4,6-Trifluoro-α-(1-methylpropyl)benzenemethanol ≥97%
2,4,6-Trifluoro-α-(1-methylpropyl)benzenemethanol

Precaución: For research use only

Formula: C₁₁H₁₃F₃O
Storage Temperature: Ambient
MDL Number: MFCD12787163
CAS Number: 1250788-98-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR