2-Fluoro-α-(1-methylpropyl)-5-(trifluoromethyl)benzenemethanol ≥97%

Supplier: AOBChem USA

Warning

214402-1G 214402-500MG 214402-5G 214402-25G
77466-904EA 284.28 USD
77466-904 77479-636 77489-922 77500-182
2-Fluoro-α-(1-methylpropyl)-5-(trifluoromethyl)benzenemethanol ≥97%
2-Fluoro-α-(1-methylpropyl)-5-(trifluoromethyl)benzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₄F₄O
MW: 250.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22906372
CAS Number: 1500943-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR