2-Chloro-6-fluoro-α-(1-methylpropyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

214390-1G 214390-500MG 214390-5G 214390-25G
77466-900EA 415.09 USD
77466-900 77479-632 77489-918 77500-178
2-Chloro-6-fluoro-α-(1-methylpropyl)benzenemethanol ≥95%
2-Chloro-6-fluoro-α-(1-methylpropyl)benzenemethanol

Precaución: For research use only

Formula: C₁₁H₁₄ClFO
MW: 216.68 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21928924
CAS Number: 1512401-31-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR