α-(1-Methylpropyl)-3-(methylthio)benzenemethanol ≥97%

Supplier: AOBChem USA

Warning

214273-1G 214273-500MG 214273-5G 214273-25G
77466-896EA 284.28 USD
77466-896 77479-628 77489-912 77500-172
α-(1-Methylpropyl)-3-(methylthio)benzenemethanol ≥97%
α-(1-Methylpropyl)-3-(methylthio)benzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₈OS
MW: 210.34 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34867572
CAS Number: 2283964-09-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR