1-Bromo-3-cyclohexylbenzene ≥95%

Supplier: AOBChem USA

Warning

214254-1G 214254-500MG 214254-5G 214254-25G
77466-892EA 383.16 USD
77466-892 77479-624 77489-908 77500-170
1-Bromo-3-cyclohexylbenzene ≥95%
1-Bromo-3-cyclohexylbenzene

Precaución: For research use only

Formula: C₁₂H₁₅Br
MW: 239.1515 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20486556
CAS Number: 19920-84-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR