1-(4-Fluoro-3-iodophenyl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

214221-1G 214221-500MG 214221-5G
77466-888EA 546.93 USD
77466-888 77479-620 77489-904
1-(4-Fluoro-3-iodophenyl)propan-1-one ≥95%
1-(4-Fluoro-3-iodophenyl)propan-1-one

Precaución: For research use only

Formula: C₉H₈FIO
MW: 278.0620 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18397048
CAS Number: 1261610-45-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR