5-Chloro-1,3-benzodioxole-4-carboxaldehyde ≥95%

Supplier: AOBChem USA

Warning

21349-1G 21349-500MG 21349-5G 21349-25G
77466-884EA 287.37 USD
77466-884 77479-616 77489-900 77500-168
5-Chloro-1,3-benzodioxole-4-carboxaldehyde ≥95%
5-Chloro-1,3-benzodioxole-4-carboxaldehyde

Precaución: For research use only

Formula: C₈H₅ClO₃
MW: 184.577 g/mol
Storage Temperature: Ambient
CAS Number: 249636-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR