2,4-Difluoro-1-propoxybenzene ≥97%

Supplier: AOBChem USA

Warning

21241-1G 21241-500MG 21241-5G 21241-25G
77466-880EA 157.59 USD
77466-880 77479-612 77489-896 77500-164
2,4-Difluoro-1-propoxybenzene ≥97%
2,4-Difluoro-1-propoxybenzene

Precaución: For research use only

Formula: C₉H₁₀F₂O
MW: 172.172 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01571111
CAS Number: 259655-00-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR