3-Bromo-5-isopropyl-2-methoxy-benzoic acid ≥97%

Supplier: AOBChem USA

Warning

21097-1G 21097-500MG 21097-5G 21097-25G
77466-876EA 315.18 USD
77466-876 77479-608 77489-892 77500-160
3-Bromo-5-isopropyl-2-methoxy-benzoic acid ≥97%
3-Bromo-5-isopropyl-2-methoxy-benzoic acid

Precaución: For research use only

Formula: C₁₁H₁₃BrO₃
MW: 273.123 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17167542
CAS Number: 1245527-83-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR