2-Fluoro-α,5-dimethyl-α-(1-methylethyl)benzenemethanol ≥96%

Supplier: AOBChem USA

Warning

210746-1G 210746-500MG 210746-5G 210746-25G
77466-872EA 415.09 USD
77466-872 77479-604 77489-888 77500-158
2-Fluoro-α,5-dimethyl-α-(1-methylethyl)benzenemethanol ≥96%
2-Fluoro-α,5-dimethyl-α-(1-methylethyl)benzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₇FO
MW: 196.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17222323
CAS Number: 1343331-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR