4-(Ethylthio)-α-methyl-α-(1-methylethyl)benzenemethanol ≥96%

Supplier: AOBChem USA

Warning

210720-1G 210720-500MG 210720-5G
77466-868EA 648.9 USD
77466-868 77479-600 77489-884
4-(Ethylthio)-α-methyl-α-(1-methylethyl)benzenemethanol ≥96%
4-(Ethylthio)-α-methyl-α-(1-methylethyl)benzenemethanol

Precaución: For research use only

Formula: C₁₃H₂₀OS
MW: 224.36 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20528538
CAS Number: 1443306-57-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR