α-Methyl-α-(1-methylethyl)-4-(2-methylpropyl)benzenemethanol ≥96%

Supplier: AOBChem USA

Warning

210705-1G 210705-500MG 210705-5G
77466-864EA 648.9 USD
77466-864 77479-596 77489-880
α-Methyl-α-(1-methylethyl)-4-(2-methylpropyl)benzenemethanol ≥96%
α-Methyl-α-(1-methylethyl)-4-(2-methylpropyl)benzenemethanol

Precaución: For research use only

Formula: C₁₅H₂₄O
MW: 220.35 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20526636
CAS Number: 1443346-65-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR