3-Ethoxy-α-methyl-α-(1-methylethyl)benzenemethanol ≥96%

Supplier: AOBChem USA

Warning

210703-1G 210703-500MG 210703-5G 210703-25G
77466-860EA 341.96 USD
77466-860 77479-592 77489-876 77500-152
3-Ethoxy-α-methyl-α-(1-methylethyl)benzenemethanol ≥96%
3-Ethoxy-α-methyl-α-(1-methylethyl)benzenemethanol

Precaución: For research use only

Formula: C₁₃H₂₀O₂
MW: 208.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20528563
CAS Number: 1443354-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR