2,6-Difluoro-α,3-Dimethyl-α-(1-methylethyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

210692-1G 210692-500MG 210692-5G
77466-856EA 447.02 USD
77466-856 77479-588 77489-872
2,6-Difluoro-α,3-Dimethyl-α-(1-methylethyl)benzenemethanol ≥95%
2,6-Difluoro-α,3-Dimethyl-α-(1-methylethyl)benzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₆F₂O
MW: 214.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24310339
CAS Number: 1557686-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR