3-Fluoro-α-methyl-α-(1-methylethyl)benzenemethanol ≥97%

Supplier: AOBChem USA

Warning

210667-1G 210667-500MG 210667-5G 210667-25G
77466-852EA 339.9 USD
77466-852 77479-584 77489-868 77500-150
3-Fluoro-α-methyl-α-(1-methylethyl)benzenemethanol ≥97%
3-Fluoro-α-methyl-α-(1-methylethyl)benzenemethanol

Precaución: For research use only

Formula: C₁₁H₁₅FO
MW: 182.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12068004
CAS Number: 37487-53-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR