4-Fluoro-α-methyl-α-(1-methylethyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

210655-1G 210655-500MG 210655-5G 210655-25G
77466-848EA 415.09 USD
77466-848 77479-580 77489-864 77500-146
4-Fluoro-α-methyl-α-(1-methylethyl)benzenemethanol ≥95%
4-Fluoro-α-methyl-α-(1-methylethyl)benzenemethanol

Precaución: For research use only

Formula: C₁₁H₁₅FO
MW: 182.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12067973
CAS Number: 28114-96-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR