α,2,5-Trimethyl-α-(1-methylethyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

210650-1G 210650-500MG 210650-5G
77466-844EA 648.9 USD
77466-844 77479-576 77489-860
α,2,5-Trimethyl-α-(1-methylethyl)benzenemethanol ≥95%
α,2,5-Trimethyl-α-(1-methylethyl)benzenemethanol

Precaución: For research use only

Formula: C₁₃H₂₀O
MW: 192.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16158863
CAS Number: 100532-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR