4-Methoxy-α-methyl-α-(1-methylethyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

210635-1G 210635-500MG 210635-5G
77466-840EA 648.9 USD
77466-840 77479-572 77489-856
4-Methoxy-α-methyl-α-(1-methylethyl)benzenemethanol ≥95%
4-Methoxy-α-methyl-α-(1-methylethyl)benzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₈O₂
MW: 194.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12068125
CAS Number: 28114-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR