3-Bromo-α-(1,1-dimethylethyl)-α-methylbenzenemethanol ≥97%

Supplier: AOBChem USA

Warning

210631-1G 210631-500MG 210631-5G 210631-25G
77466-836EA 415.09 USD
77466-836 77479-568 77489-852 77500-140
3-Bromo-α-(1,1-dimethylethyl)-α-methylbenzenemethanol ≥97%
3-Bromo-α-(1,1-dimethylethyl)-α-methylbenzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₇BrO
Storage Temperature: Ambient
MDL Number: MFCD21328246
CAS Number: 1466178-88-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR