α-(1,1-Dimethylethyl)-2-fluoro-α-methylbenzenemethanol ≥96%

Supplier: AOBChem USA

Warning

210624-1G 210624-500MG 210624-5G 210624-25G
77466-832EA 415.09 USD
77466-832 77479-564 77489-848 77500-136
α-(1,1-Dimethylethyl)-2-fluoro-α-methylbenzenemethanol ≥96%
α-(1,1-Dimethylethyl)-2-fluoro-α-methylbenzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₇FO
Storage Temperature: Ambient
MDL Number: MFCD21328241
CAS Number: 1495540-24-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR