α-(1,1-Dimethylethyl)-α-methyl[1,1'-biphenyl]-4-methanol ≥95%

Supplier: AOBChem USA

Warning

210616-1G 210616-500MG 210616-5G 210616-25G
77466-828EA 415.09 USD
77466-828 77479-560 77489-844 77500-132
α-(1,1-Dimethylethyl)-α-methyl[1,1'-biphenyl]-4-methanol ≥95%
α-(1,1-Dimethylethyl)-α-methyl[1,1'-biphenyl]-4-methanol

Precaución: For research use only

Formula: C₁₈H₂₂O
Storage Temperature: Ambient
MDL Number: MFCD23649583
CAS Number: 1546195-74-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR