α-(1,1-Dimethylethyl)-3-ethoxy-α-methylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

210600-1G 210600-500MG 210600-5G 210600-25G
77466-824EA 415.09 USD
77466-824 77479-556 77489-840 77500-128
α-(1,1-Dimethylethyl)-3-ethoxy-α-methylbenzenemethanol ≥95%
α-(1,1-Dimethylethyl)-3-ethoxy-α-methylbenzenemethanol

Precaución: For research use only

Formula: C₁₄H₂₂O₂
MW: 222.32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24198926
CAS Number: 1549021-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR