α-(1,1-Dimethylethyl)-2,6-difluoro-α,3-dimethylbenzenemethanol ≥97%

Supplier: AOBChem USA

Warning

210576-1G 210576-500MG 210576-5G 210576-25G
77466-820EA 415.09 USD
77466-820 77479-552 77489-836 77500-124
α-(1,1-Dimethylethyl)-2,6-difluoro-α,3-dimethylbenzenemethanol ≥97%
α-(1,1-Dimethylethyl)-2,6-difluoro-α,3-dimethylbenzenemethanol

Precaución: For research use only

Formula: C₁₃H₁₈F₂O
Storage Temperature: Ambient
MDL Number: MFCD24310393
CAS Number: 1556069-69-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR