3-Bromo-α-(1,1-dimethylethyl)-2-fluoro-α-methylbenzenemethanol ≥97%

Supplier: AOBChem USA

Warning

210567-1G 210567-500MG 210567-5G
77466-816EA 447.02 USD
77466-816 77479-548 77489-832
3-Bromo-α-(1,1-dimethylethyl)-2-fluoro-α-methylbenzenemethanol ≥97%
3-Bromo-α-(1,1-dimethylethyl)-2-fluoro-α-methylbenzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₆BrFO
Storage Temperature: Ambient
MDL Number: MFCD34867160
CAS Number: 1698950-49-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR