α-(1,1-Dimethylethyl)-α,3,5-trimethylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

210518-1G 210518-500MG 210518-5G 210518-25G
77466-812EA 297.67 USD
77466-812 77479-544 77489-828 77500-120
α-(1,1-Dimethylethyl)-α,3,5-trimethylbenzenemethanol ≥95%
α-(1,1-Dimethylethyl)-α,3,5-trimethylbenzenemethanol

Precaución: For research use only

Formula: C₁₄H₂₂O
MW: 206.32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24198939
CAS Number: 147611-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR