2-Chloro-6-fluoro-α-methyl-α-(1-methylethyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

210498-1G 210498-500MG 210498-5G 210498-25G
77466-808EA 226.6 USD
77466-808 77479-540 77489-824 77500-118
2-Chloro-6-fluoro-α-methyl-α-(1-methylethyl)benzenemethanol ≥95%
2-Chloro-6-fluoro-α-methyl-α-(1-methylethyl)benzenemethanol

Precaución: For research use only

Formula: C₁₁H₁₄ClFO
MW: 216.68 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29965582
CAS Number: 1872755-38-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR