α-(1,1-Dimethylethyl)-α-methylbenzenemethanol ≥97%

Supplier: AOBChem USA

Warning

210482-1G 210482-500MG 210482-5G 210482-25G
77466-804EA 415.09 USD
77466-804 77479-536 77489-820 77500-114
α-(1,1-Dimethylethyl)-α-methylbenzenemethanol ≥97%
α-(1,1-Dimethylethyl)-α-methylbenzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₈O
Storage Temperature: Ambient
MDL Number: MFCD00182590
CAS Number: 21811-48-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR