4-Ethyl-α-(1-Ethylpropyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

210471-1G 210471-500MG 210471-5G 210471-25G
77466-800EA 364.62 USD
77466-800 77479-532 77489-816 77500-110
4-Ethyl-α-(1-Ethylpropyl)benzenemethanol ≥95%
4-Ethyl-α-(1-Ethylpropyl)benzenemethanol

Precaución: For research use only

Formula: C₁₄H₂₂O
MW: 206.32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12787388
CAS Number: 1248620-17-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR