α-(1-Ethylpropyl)-4-fluorobenzenemethanol ≥97%

Supplier: AOBChem USA

Warning

210461-1G 210461-500MG 210461-5G 210461-25G
77466-796EA 297.67 USD
77466-796 77479-528 77489-812 77500-106
α-(1-Ethylpropyl)-4-fluorobenzenemethanol ≥97%
α-(1-Ethylpropyl)-4-fluorobenzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₇FO
MW: 196.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12786843
CAS Number: 1250180-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR