4-Butyl-α-(1-ethylpropyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

210451-1G 210451-500MG 210451-5G 210451-25G
77466-792EA 284.28 USD
77466-792 77479-524 77489-808 77500-102
4-Butyl-α-(1-ethylpropyl)benzenemethanol ≥95%
4-Butyl-α-(1-ethylpropyl)benzenemethanol

Precaución: For research use only

Formula: C₁₆H₂₆O
MW: 234.38 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19486857
CAS Number: 1281720-22-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR