α-(1-Ethylpropyl)-2-fluoro-5-methylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

210433-1G 210433-500MG 210433-5G 210433-25G
77466-784EA 353.29 USD
77466-784 77479-516 77489-800 77500-094
α-(1-Ethylpropyl)-2-fluoro-5-methylbenzenemethanol ≥95%
α-(1-Ethylpropyl)-2-fluoro-5-methylbenzenemethanol

Precaución: For research use only

Formula: C₁₃H₁₉FO
Storage Temperature: Ambient
MDL Number: MFCD19601784
CAS Number: 1339904-45-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR