α-(1-Ethylpropyl)-3,5-dimethylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

210428-1G 210428-500MG 210428-5G 210428-25G
77466-780EA 297.67 USD
77466-780 77479-512 77489-796 77500-090
α-(1-Ethylpropyl)-3,5-dimethylbenzenemethanol ≥95%
α-(1-Ethylpropyl)-3,5-dimethylbenzenemethanol

Precaución: For research use only

Formula: C₁₄H₂₂O
Storage Temperature: Ambient
MDL Number: MFCD19601782
CAS Number: 1341801-91-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR