α-(1-Ethylpropyl)-3-fluorobenzenemethanol ≥97%

Supplier: AOBChem USA

Warning

210419-1G 210419-500MG 210419-5G 210419-25G
77466-776EA 297.67 USD
77466-776 77479-508 77489-792 77500-086
α-(1-Ethylpropyl)-3-fluorobenzenemethanol ≥97%
α-(1-Ethylpropyl)-3-fluorobenzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₇FO
MW: 196.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19601798
CAS Number: 1343273-52-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR