3-Ethoxy-α-(1-ethylpropyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

210395-1G 210395-500MG 210395-5G 210395-25G
77466-772EA 415.09 USD
77466-772 77479-504 77489-788 77500-082
3-Ethoxy-α-(1-ethylpropyl)benzenemethanol ≥95%
3-Ethoxy-α-(1-ethylpropyl)benzenemethanol

Precaución: For research use only

Formula: C₁₄H₂₂O₂
MW: 222.32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23833747
CAS Number: 1504863-84-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR