α-(1-Ethylpropyl)-2-fluoro-3-(trifluoromethyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

210392-1G 210392-500MG 210392-5G 210392-25G
77466-768EA 415.09 USD
77466-768 77479-500 77489-784 77500-080
α-(1-Ethylpropyl)-2-fluoro-3-(trifluoromethyl)benzenemethanol ≥95%
α-(1-Ethylpropyl)-2-fluoro-3-(trifluoromethyl)benzenemethanol

Precaución: For research use only

Formula: C₁₃H₁₆F₄O
MW: 264.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22912891
CAS Number: 1505755-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR