α-(1-Ethylpropyl)-3,5-dimethoxybenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

210364-1G 210364-500MG 210364-5G 210364-25G
77466-764EA 415.09 USD
77466-764 77479-496 77489-780 77500-076
α-(1-Ethylpropyl)-3,5-dimethoxybenzenemethanol ≥95%
α-(1-Ethylpropyl)-3,5-dimethoxybenzenemethanol

Precaución: For research use only

Formula: C₁₄H₂₂O₃
Storage Temperature: Ambient
MDL Number: MFCD22910313
CAS Number: 1538021-34-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR