5-Bromo-α-(1-ethylpropyl)-2-fluorobenzenemethanol ≥97%

Supplier: AOBChem USA

Warning

210357-1G 210357-500MG 210357-5G 210357-25G
77466-760EA 297.67 USD
77466-760 77479-492 77489-776 77500-072
5-Bromo-α-(1-ethylpropyl)-2-fluorobenzenemethanol ≥97%
5-Bromo-α-(1-ethylpropyl)-2-fluorobenzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₆BrFO
MW: 275.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23759108
CAS Number: 1540868-45-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR