α-(1-Ethylpropyl)-4-methoxy-3,5-dimethylbenzenemethanol ≥97%

Supplier: AOBChem USA

Warning

210350-1G 210350-500MG 210350-5G 210350-25G
77466-756EA 312.09 USD
77466-756 77479-488 77489-772 77500-070
α-(1-Ethylpropyl)-4-methoxy-3,5-dimethylbenzenemethanol ≥97%
α-(1-Ethylpropyl)-4-methoxy-3,5-dimethylbenzenemethanol

Precaución: For research use only

Formula: C₁₅H₂₄O₂
Storage Temperature: Ambient
MDL Number: MFCD25296963
CAS Number: 1547213-63-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR