3-(Dimethylamino)-α-(1-ethylpropyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

210344-1G 210344-500MG 210344-5G 210344-25G
77466-752EA 415.09 USD
77466-752 77479-482 77489-766 77500-066
3-(Dimethylamino)-α-(1-ethylpropyl)benzenemethanol ≥95%
3-(Dimethylamino)-α-(1-ethylpropyl)benzenemethanol

Precaución: For research use only

Formula: C₁₄H₂₃NO
MW: 221.34 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34867105
CAS Number: 1592252-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR