5-Chloro-2,3-difluoro-4-pyridinecarboxylic acid ≥97%

Supplier: AOBChem USA

Warning

21034-1G 21034-500MG 21034-5G 21034-25G
77466-748EA 151.41 USD
77466-748 77479-484 77489-768 77500-064
5-Chloro-2,3-difluoro-4-pyridinecarboxylic acid ≥97%
5-Chloro-2,3-difluoro-4-pyridinecarboxylic acid

Precaución: For research use only

Formula: C₆H₂ClF₂NO₂
MW: 193.535 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15528689
CAS Number: 406676-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR