5-Chloro-2,3-difluoropyridine-4-boronic acid ≥97%

Supplier: AOBChem USA

Warning

21032-1G 21032-500MG 21032-5G 21032-25G
77466-744EA 399.64 USD
77466-744 77479-476 77489-760 77500-058
5-Chloro-2,3-difluoropyridine-4-boronic acid ≥97%
5-Chloro-2,3-difluoropyridine-4-boronic acid

Precaución: For research use only

Formula: C₅H₃BClF₂NO₂
MW: 193.344 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18382789
CAS Number: 1239491-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR